Learn More
Medchemexpress LLC HY-18632A 5mg , GSK2879552 (dihydrochloride) CAS:1902123-72-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18632A5MG
Encompass_Preferred
HY-18632A 5mg GSK2879552 (dihydrochloride) CAS: 1902123-72-1 GSK2879552 dihydrochloride an orally active, selective and irreversible inhibitor of lysine specific demethylase 1 (LSD1/KDM1A), with potential antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1506