Learn More
Medchemexpress LLC GSK2879552 dihydrochloride | 1902123-72-1 | 99.8% | 437.40 | 100 MG
SDP
Catalog No. 5000387813
Encompass_Preferred
GSK2879552 dihydrochloride is an orally active, selective, and irreversible inhibitor of lysine specific demethylase 1 (LSD1/KDM1A), which has potential antineoplastic activity. For research use only, it is not sold to patients.
  • Orally active.
  • Selective and irreversible inhibitor of LSD1/KDM1A.
  • Exhibits potential antineoplastic activity.
  • Inhibits KDM1A histone demethylase activity, inducing differentiation of sorafenib-resistant cells and attenuating stemness properties in vitro.
  • Depresses the transcription of Wnt antagonists and downregulates β-catenin signaling activity in sorafenib-resistant cells in vitro.
  • In vivo treatment with 1.5 mg/kg exhibits tumor growth inhibition in SCLC xenograft bearing mice.
Catalog No. 50-003-87813 Supplier Medchemexpress LLC Supplier No. HY18632A100MG
GSK2879552 dihydrochloride is an orally active, selective, and irreversible inhibitor of lysine specific demethylase 1 (LSD1/KDM1A), which has potential antineoplastic activity. For research use only, it is not sold to patients.
  • Orally active.
  • Selective and irreversible inhibitor of LSD1/KDM1A.
  • Exhibits potential antineoplastic activity.
  • Inhibits KDM1A histone demethylase activity, inducing differentiation of sorafenib-resistant cells and attenuating stemness properties in vitro.
  • Depresses the transcription of Wnt antagonists and downregulates β-catenin signaling activity in sorafenib-resistant cells in vitro.
  • In vivo treatment with 1.5 mg/kg exhibits tumor growth inhibition in SCLC xenograft bearing mice.