Learn More
Medchemexpress LLC GSK2879552 dihydrochloride | 1902123-72-1 | MFCD30481342 | 99.8% | 437.4 g·mol⁻1 | C23H30Cl2N2O2 | 10 MG
SDP
Catalog No. 5000298605
Encompass_Preferred
GSK2879552 dihydrochloride is the dihydrochloride salt of an orally bioavailable, selective, mechanism-based irreversible inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A). It is used as a research reagent in epigenetic and oncology studies and has been evaluated in early clinical trials for hematologic and solid tumor indications.
  • Dihydrochloride salt form for improved solubility.
  • Irreversible LSD1 inhibitor (mechanism-based inactivator).
  • Orally bioavailable scaffold used in preclinical and clinical research.
  • High purity (≈99.8%) suitable for research applications.
  • Available in multiple solid and solution formats and milligram quantities.
Catalog No. 50-002-98605 Supplier Medchemexpress LLC Supplier No. HY18632A10MG
GSK2879552 dihydrochloride is the dihydrochloride salt of an orally bioavailable, selective, mechanism-based irreversible inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A). It is used as a research reagent in epigenetic and oncology studies and has been evaluated in early clinical trials for hematologic and solid tumor indications.
  • Dihydrochloride salt form for improved solubility.
  • Irreversible LSD1 inhibitor (mechanism-based inactivator).
  • Orally bioavailable scaffold used in preclinical and clinical research.
  • High purity (≈99.8%) suitable for research applications.
  • Available in multiple solid and solution formats and milligram quantities.