Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK2945 hydrochloride | 00-00-0 | 99.7% | 457.80 g/mol | C20H19Cl3N2O2S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY117147A5MG

Encompass_Preferred

GSK2945 hydrochloride is the hydrochloride salt of a small-molecule antagonist of the nuclear receptor Rev-erbα, supplied for preclinical research. It has reported EC50 values of 21.5 μM (mouse) and 20.8 μM (human) and has been used to increase CYP7A1 levels and modulate cholesterol metabolism.

  • High chemical purity (99.7%).
  • Solid form suitable for storage at 4°C; in solution store under nitrogen at -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO (~20.83 mg/mL); in vivo formulations and protocols are available for suspension or clear solution preparations.
  • Activity: Rev-erbα antagonist with reported EC50 around 21 μM in cell-based assays.
  • Provided for research use with supporting assay and formulation data.

Catalog No. 50-002-64578


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.