Learn More
Medchemexpress LLC GSK2945 (hydrochloride) | 2517771-89-8 | 98.5% | 457.80 | 1 ML
SDP
Catalog No. 5000436804
Encompass_Preferred
GSK2945 hydrochloride is a tertiary amine and a highly specific Rev-erbα/REV-ERBα (mouse/human reverse erythroblastosis virus α) antagonist with EC50s of 21.5 μM and 20.8 μM, respectively. It enhances cholesterol 7α-hydroxylase (CYP7A1) levels and cholesterol metabolism. For research use only.
  • Highly specific Rev-erbα/REV-ERBα antagonist.
  • Enhances cholesterol 7α-hydroxylase (CYP7A1) levels.
  • Promotes cholesterol metabolism.
  • Suitable for research applications.
Catalog No. 50-004-36804 Supplier Medchemexpress LLC Supplier No. HY117147A10MM/1ML
GSK2945 hydrochloride is a tertiary amine and a highly specific Rev-erbα/REV-ERBα (mouse/human reverse erythroblastosis virus α) antagonist with EC50s of 21.5 μM and 20.8 μM, respectively. It enhances cholesterol 7α-hydroxylase (CYP7A1) levels and cholesterol metabolism. For research use only.
  • Highly specific Rev-erbα/REV-ERBα antagonist.
  • Enhances cholesterol 7α-hydroxylase (CYP7A1) levels.
  • Promotes cholesterol metabolism.
  • Suitable for research applications.