Learn More
Medchemexpress LLC GSK2945 (hydrochloride) | 98.5% | 457.80 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY117147A50MG
Encompass_Preferred
GSK2945 hydrochloride is a tertiary amine and a highly specific Rev-erbα/REV-ERBα (mouse/human reverse erythroblastosis virus α) antagonist. It has EC50s of 21.5 μM and 20.8 μM, respectively. This compound functions by enhancing cholesterol 7α-hydroxylase (CYP7A1) levels and promoting cholesterol metabolism.
  • Highly specific antagonist.
  • Enhances cholesterol 7α-hydroxylase levels.
  • Promotes cholesterol metabolism.
Catalog No. 50-004-36291