Learn More
Selleck Chemical LLC GSK2982772 2mg 1622848-92-3
SDP
Catalog No. 501948091
Encompass_Preferred
GSK2982772 is an ATP competitive receptor-interacting protein-1 (RIP1) kinase (RIPK1) inhibitor with the IC50 value of 16 nM. It has exquisite kinase specificity and excellent activity in blocking many TNF-dependent cellular responses. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8091 Supplier Selleck Chemical LLC Supplier No. S84842MG
GSK2982772 is an ATP competitive receptor-interacting protein-1 (RIP1) kinase (RIPK1) inhibitor with the IC50 value of 16 nM. It has exquisite kinase specificity and excellent activity in blocking many TNF-dependent cellular responses. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.