Learn More
Medchemexpress LLC HY-101760 5mg , GSK2982772 CAS:1622848-92-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1017605MG
Encompass_Preferred
HY-101760 5mg GSK2982772 CAS: 1622848-92-3 GSK2982772 is a potent, orally active and ATP competitive RIP1 kinase inhibitor with IC50 values of 16 nM and 20 nM for human and monkey RIP1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9426