Learn More
Medchemexpress LLC Gsk2983559 | 99.5% | 612.63 | 100 MG
SDP
Catalog No. 5000438927
Encompass_Preferred
GSK2983559 is an orally active and potent receptor interacting protein 2 (RIP2) kinase inhibitor. It blocks many proinflammatory cytokine responses both in vivo and in human inflammatory bowel disease explant samples.
Catalog No. 50-004-38927 Supplier Medchemexpress LLC Supplier No. HY112038A100MG
GSK2983559 is an orally active and potent receptor interacting protein 2 (RIP2) kinase inhibitor. It blocks many proinflammatory cytokine responses both in vivo and in human inflammatory bowel disease explant samples.