Learn More
TARGETMOL CHEMICALS INC GSK2983559 ACTIVE METABO 10MG
SDP
Catalog No. 502579775
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2983559 active metabolite (RIPK2-IN-1) is a potent selective receptor-interacting protein-2 (RIP2) kinase inhibitor. purity: 98%
Catalog No. 50-257-9775 Supplier TARGETMOL CHEMICALS INC Supplier No. T540110MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2983559 active metabolite (RIPK2-IN-1) is a potent selective receptor-interacting protein-2 (RIP2) kinase inhibitor. purity: 98%