Learn More
Medchemexpress LLC HY-112038 5mg Medchemexpress, GSK2983559 (free acid) CAS:1579965-12-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1120385MG
Encompass_Preferred
Medchemexpress, HY-112038 5mg GSK2983559 (free acid) CAS:1579965-12-0 GSK2983559 free acid (compound 3) is a potent, specific and oral bioavailable receptor interacting protein 2 (RIP2) kinase inhibitor. GSK2983559 free acid has excellent activity in blocking many proinflammatory cytokine responses in vivo and in human inflammatory bowel disease explant samples. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8995