Learn More
TARGETMOL CHEMICALS INC GSK299115A 5MG
SDP
Catalog No. 502580608
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK299115A (GSK-299115A) is a selective ROCK1 Inhibitor. GSK299115A exhibits IC50 of 8nM 620nM 560nM for ROCK1 RSK1 p70S6K respectively. purity: 99%
Catalog No. 50-258-0608 Supplier TARGETMOL CHEMICALS INC Supplier No. T274715MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK299115A (GSK-299115A) is a selective ROCK1 Inhibitor. GSK299115A exhibits IC50 of 8nM 620nM 560nM for ROCK1 RSK1 p70S6K respectively. purity: 99%