Learn More
Enzo Life Sciences GSK3 a/b (pTyr279/216) monoclonal antibody (6D3) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905762100
Encompass_Preferred
Clone: 6D3. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of mouse GSK phosphorylated at Tyr216/279. UniProt ID: Q2NL51 (GSK3A), Q9WV60 (GSK3B). Species reactivity: Human, Mouse. Applications: WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -80°C.
Catalog No. 50-200-7040