Learn More
Medchemexpress LLC Gsk3β inhibitor II | 478482-75-6 | 99.5% | 395.22 | 25 MG
SDP
Catalog No. 5000363465
Encompass_Preferred
Gsk3β inhibitor II is an inhibitor of Gsk3β and can be used for research related to Alzheimer's disease (AD). It demonstrates specific interactions within biological systems, contributing to its inhibitory action.
  • Forms hydrogen bonds with Val135
  • Exhibits hydrophobic interaction with Ile62, Phe67, Val70, Ala83, and Leu188
  • Shows ionic interaction with Lys85
  • Can be used for Alzheimer's disease research
Catalog No. 50-003-63465 Supplier Medchemexpress LLC Supplier No. 25MGHY14679
Gsk3β inhibitor II is an inhibitor of Gsk3β and can be used for research related to Alzheimer's disease (AD). It demonstrates specific interactions within biological systems, contributing to its inhibitory action.
  • Forms hydrogen bonds with Val135
  • Exhibits hydrophobic interaction with Ile62, Phe67, Val70, Ala83, and Leu188
  • Shows ionic interaction with Lys85
  • Can be used for Alzheimer's disease research