Learn More
Cayman Chemical GSK3 InhIbItr II 5mg
SDP
Catalog No. 502983433
Encompass_Preferred
A potent inhibitor of GSK3β (IC50 = 390 nM) that does not inhibit GSK-3α
Catalog No. 50-298-3433 Supplier Cayman Chemical Supplier No. 190775 MG
A potent inhibitor of GSK3β (IC50 = 390 nM) that does not inhibit GSK-3α