Learn More
Cayman Chemical GSK3 InhIbItr XI 1mg
SDP
Catalog No. 502994417
Encompass_Preferred
A potent inhibitor of GSK3β (Ki = 25 nM); inhibits GSK3β in HEK293 cells (EC50 = 32 nM)
Catalog No. 50-299-4417 Supplier Cayman Chemical Supplier No. 239401 MG
A potent inhibitor of GSK3β (Ki = 25 nM); inhibits GSK3β in HEK293 cells (EC50 = 32 nM)