Learn More
Medchemexpress LLC HY-100459 1mg Medchemexpress, GSK3179106 CAS:1627856-64-7 Purity:>98%
SDP
Catalog No. 501931844
Encompass_Preferred
Medchemexpress, HY-100459 1mg GSK3179106 CAS:1627856-64-7 GSK3179106 is a potent and selective RET kinase inhibitor with an IC 50 of 0.4 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1844 Supplier Medchemexpress LLC Supplier No. HY1004591MG
Medchemexpress, HY-100459 1mg GSK3179106 CAS:1627856-64-7 GSK3179106 is a potent and selective RET kinase inhibitor with an IC 50 of 0.4 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.