Learn More
Selleck Chemical LLC GSK3179106 2mg 1627856-64-7
SDP
Supplier:  Selleck Chemical LLC S88212MG
Encompass_Preferred
GSK3179106 is a potent, selective, and gut-restricted pyridone hinge binder small molecule RET (c-RET) kinase inhibitor with a RET IC50 of 0.3 nM and is efficacious in vivo. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8412