Learn More
Medchemexpress LLC GSK3182571 | 2135595-04-7 | 99.8% | 495.02 | C25H31ClN8O | 25MG
SDP
Catalog No. 5000236399
Encompass_Preferred
GSK3182571 is a small-molecule pan-kinase inhibitor used for laboratory research to probe kinase signaling and assess off-target kinase effects in biochemical and cellular assays. The compound is supplied at high purity and is typically handled in DMSO; it is intended for research use only and not for human or clinical applications.
  • Pan-kinase inhibitor for broad kinase profiling
  • High purity suitable for biochemical and cellular assays
  • Available in small research pack sizes
  • Soluble in DMSO for assay preparation
  • Intended for research use; not for clinical use
Catalog No. 50-002-36399 Supplier Medchemexpress LLC Supplier No. HY1240025MG
GSK3182571 is a small-molecule pan-kinase inhibitor used for laboratory research to probe kinase signaling and assess off-target kinase effects in biochemical and cellular assays. The compound is supplied at high purity and is typically handled in DMSO; it is intended for research use only and not for human or clinical applications.
  • Pan-kinase inhibitor for broad kinase profiling
  • High purity suitable for biochemical and cellular assays
  • Available in small research pack sizes
  • Soluble in DMSO for assay preparation
  • Intended for research use; not for clinical use