Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK3182571 | 2135595-04-7 | 99.8% | 495.02 g/mol | C25H31ClN8O | 100MG
SDP

Catalog No. 5000236709
Encompass_Preferred

GSK3182571 is a small-molecule pan-kinase inhibitor for research use that modulates Akt signaling and the PI3K/Akt/mTOR pathway. It is provided as a 100 mg research sample intended for in vitro pharmacology and biochemical studies, including probing kinase networks and off-target effects in cellular models. The compound is typically stored at -20°C and is soluble in DMSO.

  • Pan-kinase inhibitor active against Akt and related kinases.
  • High purity suitable for research assays.
  • Molecular formula C25H31ClN8O; molecular weight 495.02 g/mol.
  • Soluble in DMSO for assay preparation.
  • Intended for in vitro pharmacology and biochemical studies.
  • Store at -20°C to maintain stability.
  • Supplied as a 100 mg research sample.

Catalog No. 50-002-36709 Supplier Medchemexpress LLC Supplier No. HY12400100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK3182571 is a small-molecule pan-kinase inhibitor for research use that modulates Akt signaling and the PI3K/Akt/mTOR pathway. It is provided as a 100 mg research sample intended for in vitro pharmacology and biochemical studies, including probing kinase networks and off-target effects in cellular models. The compound is typically stored at -20°C and is soluble in DMSO.

  • Pan-kinase inhibitor active against Akt and related kinases.
  • High purity suitable for research assays.
  • Molecular formula C25H31ClN8O; molecular weight 495.02 g/mol.
  • Soluble in DMSO for assay preparation.
  • Intended for in vitro pharmacology and biochemical studies.
  • Store at -20°C to maintain stability.
  • Supplied as a 100 mg research sample.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.