Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK3182571 | 2135595-04-7 | 99.8% | 495.02 g/mol | C25H31ClN8O | 50MG
SDP

Catalog No. 5000236953
Encompass_Preferred

GSK3182571 is a non-selective, broad-spectrum pan-kinase inhibitor provided as a solid powder for research use. It is intended for in vitro and cell-based studies to probe kinase networks and off-target effects; not for human use.

  • High purity (99.75%).
  • Molecular weight 495.02 g/mol.
  • Molecular formula C25H31ClN8O.
  • Soluble in DMSO at 100 mg/mL (202.01 mM) with ultrasonic aid and pH adjustment.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • White to light yellow solid appearance.

Catalog No. 50-002-36953 Supplier Medchemexpress LLC Supplier No. HY1240050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK3182571 is a non-selective, broad-spectrum pan-kinase inhibitor provided as a solid powder for research use. It is intended for in vitro and cell-based studies to probe kinase networks and off-target effects; not for human use.

  • High purity (99.75%).
  • Molecular weight 495.02 g/mol.
  • Molecular formula C25H31ClN8O.
  • Soluble in DMSO at 100 mg/mL (202.01 mM) with ultrasonic aid and pH adjustment.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • White to light yellow solid appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.