Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK3182571 | 2135595-04-7 | MFCD28137930 | 99.8% | 495.02 | C25H31ClN8O | 10MG
SDP

Catalog No. 5000238990
Encompass_Preferred

GSK3182571 is a non-selective, broad-spectrum pan-kinase inhibitor used in research to probe kinase networks and assess off-target effects; it can induce changes in target protein thermostability in cellular assays.

  • Pan-kinase inhibitor for kinase network and off-target studies.
  • High purity: 99.8%.
  • Molecular weight 495.02 g/mol; chemical formula C25H31ClN8O.
  • Soluble in DMSO at 100 mg/mL (202.01 mM); use ultrasonication and adjust pH to 4 if required.
  • Store powder at -20°C (3 years) or 4°C (2 years); store solutions at -80°C for long-term stability.
  • Packaged in small milligram quantities suitable for assay development and biochemical testing.

Catalog No. 50-002-38990 Supplier Medchemexpress LLC Supplier No. HY1240010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK3182571 is a non-selective, broad-spectrum pan-kinase inhibitor used in research to probe kinase networks and assess off-target effects; it can induce changes in target protein thermostability in cellular assays.

  • Pan-kinase inhibitor for kinase network and off-target studies.
  • High purity: 99.8%.
  • Molecular weight 495.02 g/mol; chemical formula C25H31ClN8O.
  • Soluble in DMSO at 100 mg/mL (202.01 mM); use ultrasonication and adjust pH to 4 if required.
  • Store powder at -20°C (3 years) or 4°C (2 years); store solutions at -80°C for long-term stability.
  • Packaged in small milligram quantities suitable for assay development and biochemical testing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.