Learn More
Medchemexpress LLC HY-101563 10mg Medchemexpress, GSK3326595 CAS:1616392-22-3 Purity:>98%
SDP
Catalog No. 502028667
Encompass_Preferred
Medchemexpress, HY-101563 10mg GSK3326595 CAS:1616392-22-3 GSK3326595 (EPZ015938) is a potent, selective, reversible inhibitor of protein arginine methyltransferase 5 (PRMT5) with an IC50 of 6.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8667 Supplier Medchemexpress LLC Supplier No. HY10156310MG
Medchemexpress, HY-101563 10mg GSK3326595 CAS:1616392-22-3 GSK3326595 (EPZ015938) is a potent, selective, reversible inhibitor of protein arginine methyltransferase 5 (PRMT5) with an IC50 of 6.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.