Learn More
Medchemexpress LLC GSK3368715 | 1629013-22-4 | 99.4% | 10 MG
SDP
Catalog No. 5000341069
Encompass_Preferred
GSK3368715 is an orally active, reversible type I protein arginine methyltransferases (PRMTs) inhibitor. It induces a shift in arginine methylation states, modifies exon usage, and exhibits strong anti-cancer activity, making it suitable for oncology research.
  • Orally active and reversible inhibitor
  • Inhibits type I protein arginine methyltransferases (PRMTs)
  • Induces shifts in arginine methylation states
  • Modifies exon usage
  • Demonstrates strong anti-cancer activity
  • Effective against a broad range of cancer cell lines
  • Target: Histone Methyltransferase
  • Pathway: Epigenetics
Catalog No. 50-003-41069 Supplier Medchemexpress LLC Supplier No. HY12871710MG
GSK3368715 is an orally active, reversible type I protein arginine methyltransferases (PRMTs) inhibitor. It induces a shift in arginine methylation states, modifies exon usage, and exhibits strong anti-cancer activity, making it suitable for oncology research.
  • Orally active and reversible inhibitor
  • Inhibits type I protein arginine methyltransferases (PRMTs)
  • Induces shifts in arginine methylation states
  • Modifies exon usage
  • Demonstrates strong anti-cancer activity
  • Effective against a broad range of cancer cell lines
  • Target: Histone Methyltransferase
  • Pathway: Epigenetics