Learn More
Medchemexpress LLC HY-128717A 10mg Medchemexpress, GSK3368715 (dihydrochloride) CAS:1628925-77-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128717A10MG
Encompass_Preferred
Medchemexpress, HY-128717A 10mg GSK3368715 (dihydrochloride) CAS:1628925-77-8 GSK3368715 dihydrochloride (EPZ019997 dihydrochloride) is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor (IC50=3.1 nM (PRMT1), 48 nM (PRMT3), 1148 nM (PRMT4), 5.7 nM (PRMT6), 1.7 nM (PRMT8)). GSK3368715 dihydrochloride (EPZ019997 dihydrochloride) produces a shift in arginine methylation states, alters exon usage, and has strong anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8926