Learn More
Medchemexpress LLC Gsk3368715 dihydrochloride | 1628925-77-8 | 100.0% | 25 MG
SDP
Catalog No. 5000341632
Encompass_Preferred
GSK3368715 dihydrochloride (EPZ019997 dihydrochloride) is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor. It produces a shift in arginine methylation states, alters exon usage, and exhibits strong anti-cancer activity. This compound shows 50% or more growth inhibition relative to DMSO-treated cells in the majority of 249 cancer cell lines, representing 12 tumor types. In vivo, it significantly affects the growth of BxPC3 xenografts, reducing tumor growth by 78% and 97% in the 150- and 300-mg/kg dose groups, respectively.
  • Orally active
  • Reversible inhibitor
  • SAM uncompetitive type I PRMTs inhibitor
  • Shifts arginine methylation states
  • Alters exon usage
  • Exhibits strong anti-cancer activity
  • Inhibits growth in various cancer cell lines
  • Reduces tumor growth in BxPC3 xenografts
Catalog No. 50-003-41632 Supplier Medchemexpress LLC Supplier No. 25MGHY128717A
GSK3368715 dihydrochloride (EPZ019997 dihydrochloride) is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor. It produces a shift in arginine methylation states, alters exon usage, and exhibits strong anti-cancer activity. This compound shows 50% or more growth inhibition relative to DMSO-treated cells in the majority of 249 cancer cell lines, representing 12 tumor types. In vivo, it significantly affects the growth of BxPC3 xenografts, reducing tumor growth by 78% and 97% in the 150- and 300-mg/kg dose groups, respectively.
  • Orally active
  • Reversible inhibitor
  • SAM uncompetitive type I PRMTs inhibitor
  • Shifts arginine methylation states
  • Alters exon usage
  • Exhibits strong anti-cancer activity
  • Inhibits growth in various cancer cell lines
  • Reduces tumor growth in BxPC3 xenografts