Learn More
Medchemexpress LLC Gsk3368715 dihydrochloride | 1628925-77-8 | 99.96% | 100 MG
SDP
Catalog No. 5000341277
Encompass_Preferred
GSK3368715 dihydrochloride is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor. It exhibits potent anti-cancer activity by causing a shift in arginine methylation states and modifying exon usage.
  • Orally active and reversible
  • Inhibits type I protein arginine methyltransferases (PRMTs)
  • Demonstrates strong anti-cancer activity
  • Modifies exon usage
  • Causes a shift in arginine methylation states
Catalog No. 50-003-41277 Supplier Medchemexpress LLC Supplier No. HY128717A100MG
GSK3368715 dihydrochloride is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor. It exhibits potent anti-cancer activity by causing a shift in arginine methylation states and modifying exon usage.
  • Orally active and reversible
  • Inhibits type I protein arginine methyltransferases (PRMTs)
  • Demonstrates strong anti-cancer activity
  • Modifies exon usage
  • Causes a shift in arginine methylation states