Learn More
Medchemexpress LLC GSK3368715 (EPZ019997) | 1629013-22-4 | 99.4% | 50 MG
SDP
Catalog No. 5000342321
Encompass_Preferred
GSK3368715 (EPZ019997) is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor. It shifts arginine methylation states, alters exon usage, and possesses strong anti-cancer activity.
  • Orally active and reversible PRMT inhibitor
  • Inhibits PRMT1 (IC50 3.1 nM), PRMT3 (IC50 48 nM), PRMT6 (IC50 5.7 nM), and PRMT8 (IC50 1.7 nM)
  • Shows 50% or more growth inhibition in the majority of 249 cancer cell lines
  • Significantly affects growth of BxPC3 xenografts, reducing tumor growth by up to 97%
Catalog No. 50-003-42321 Supplier Medchemexpress LLC Supplier No. HY12871750MG
GSK3368715 (EPZ019997) is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor. It shifts arginine methylation states, alters exon usage, and possesses strong anti-cancer activity.
  • Orally active and reversible PRMT inhibitor
  • Inhibits PRMT1 (IC50 3.1 nM), PRMT3 (IC50 48 nM), PRMT6 (IC50 5.7 nM), and PRMT8 (IC50 1.7 nM)
  • Shows 50% or more growth inhibition in the majority of 249 cancer cell lines
  • Significantly affects growth of BxPC3 xenografts, reducing tumor growth by up to 97%