Learn More
Selleck Chemical LLC GSK33687153HCl 5mg
SDP
Supplier:  Selleck Chemical LLC S88585MG
Encompass_Preferred
GSK3368715 is a potent inhibitor of type I protein arginine methyltransferases (PRMT) that inhibits PRMT1, 3, 4, 6 and 8 with Kiapp vaules ranging from 1.5 to 81 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3406