Learn More
Selleck Chemical LLC GSK343 5mg 1346704-33-3
SDP
Catalog No. NC1078466
Encompass_Preferred
GSK343 is a potent and selective EZH2 inhibitor with IC50 of 4 nM in a cell-free assay, showing 60 fold selectivity against EZH1, and >1000 fold selectivity against other histone methyltransferases. GSK343 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1078466 Supplier Selleck Chemical LLC Supplier No. S71645MG
GSK343 is a potent and selective EZH2 inhibitor with IC50 of 4 nM in a cell-free assay, showing 60 fold selectivity against EZH1, and >1000 fold selectivity against other histone methyltransferases. GSK343 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.