Learn More
Selleck Chemical LLC GSK3685032-E1046-5MG
SDP
Catalog No. 503134426
Encompass_Preferred
GSK3685032 is a non-time-dependent highly selective enzymatic inhibitor of DNA methyltransferase (DNMT1) with IC50 of 0 036 M and over 2500-fold more selective than DNMT3A/3L and DNMT3B/3L
Catalog No. 50-313-4426 Supplier Selleck Chemical LLC Supplier No. E10465MG
GSK3685032 is a non-time-dependent highly selective enzymatic inhibitor of DNA methyltransferase (DNMT1) with IC50 of 0 036 M and over 2500-fold more selective than DNMT3A/3L and DNMT3B/3L