Learn More
Medchemexpress LLC HY-12082A 2mg Medchemexpress, GSK369796 Dihydrochloride CAS:1010411-21-8 Purity:>98%
SDP
Catalog No. 502080953
Encompass_Preferred
Medchemexpress, HY-12082A 2mg GSK369796 Dihydrochloride CAS:1010411-21-8 GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0953 Supplier Medchemexpress LLC Supplier No. HY12082A2MG
Medchemexpress, HY-12082A 2mg GSK369796 Dihydrochloride CAS:1010411-21-8 GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.