Learn More
Cayman Chemical GSK369796hydrochlorIde 10mg
SDP
Catalog No. 502912639
Encompass_Preferred
An antimalarial agent; active against the chloroquine-sensitive P. falciparum strains 3D7 and HB3 (IC50s = 11.2 and 12.6 nM, respectively) and the chloroquine-resistant strain K1 (IC50 = 13.2 nM); reduces parasitemia in mice infected with P. berghei (ED50 = 2.8 mg/kg)
Catalog No. 50-291-2639 Supplier Cayman Chemical Supplier No. 3606110 MG
An antimalarial agent; active against the chloroquine-sensitive P. falciparum strains 3D7 and HB3 (IC50s = 11.2 and 12.6 nM, respectively) and the chloroquine-resistant strain K1 (IC50 = 13.2 nM); reduces parasitemia in mice infected with P. berghei (ED50 = 2.8 mg/kg)