Learn More
Medchemexpress LLC HY-101746 5mg Medchemexpress, GSK376501A CAS:1010412-80-2 Purity:>98%
SDP
Catalog No. 502080642
Encompass_Preferred
Medchemexpress, HY-101746 5mg GSK376501A CAS:1010412-80-2 GSK376501A is a selective peroxisome proliferator-activated receptor gamma (PPARγ) modulator for the treatment of type 2 diabetes mellitus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0642 Supplier Medchemexpress LLC Supplier No. HY1017465MG
Medchemexpress, HY-101746 5mg GSK376501A CAS:1010412-80-2 GSK376501A is a selective peroxisome proliferator-activated receptor gamma (PPARγ) modulator for the treatment of type 2 diabetes mellitus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.