Learn More
Apexbio Technology LLC GSK3787, 10mg. Cas: 188591-46-0 MFCD: MFCD00099612. PPARβ/δ antagonist,novel and irreversible
SDP
Supplier:  Apexbio Technology LLC A430310
Encompass_Preferred
GSK3787 is a novel and irreversible antagonist of the peroxisome proliferator-activated receptor-/ (PPAR-/) that covalently binds to a cysteine residue in the ligand-binding domain of PPAR- /. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4911