Learn More
Apexbio Technology LLC GSK3787 188591-46-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062659
Encompass_Preferred
GSK3787 is an irreversible antagonist targeting peroxisome proliferator-activated receptor beta/delta (PPAR- / ) Structurally it acts through covalent modification by binding specific cysteine residues within the ligand-binding domain of PPAR- / thereby disrupting receptor activation and subsequent co-regulator interactions Due to receptor selectivity GSK3787 preferentially inhibits PPAR- / relative to PPAR- Experimental evidence in rodent models demonstrates that oral administration of GSK3787 counteracts agonist-induced gene expression such as Angptl4 and Adrp by reducing recruitment of activated PPAR- / to target gene promoters indicating potential utility in studying PPAR- / -mediated regulatory pathways related to lipid metabolism and gene transcription in vitro and in vivo
Catalog No. 50-000-62659 Supplier Apexbio Technology LLC Supplier No. A43035.1
GSK3787 is an irreversible antagonist targeting peroxisome proliferator-activated receptor beta/delta (PPAR- / ) Structurally it acts through covalent modification by binding specific cysteine residues within the ligand-binding domain of PPAR- / thereby disrupting receptor activation and subsequent co-regulator interactions Due to receptor selectivity GSK3787 preferentially inhibits PPAR- / relative to PPAR- Experimental evidence in rodent models demonstrates that oral administration of GSK3787 counteracts agonist-induced gene expression such as Angptl4 and Adrp by reducing recruitment of activated PPAR- / to target gene promoters indicating potential utility in studying PPAR- / -mediated regulatory pathways related to lipid metabolism and gene transcription in vitro and in vivo