Learn More
TARGETMOL CHEMICALS INC GSK3787 25MG
SDP
Catalog No. 502581049
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK3787 is a specific and irreversible PPAR(delta) inhibitor(pIC50= 6.6). purity: 98%
Catalog No. 50-258-1049 Supplier TARGETMOL CHEMICALS INC Supplier No. T194125MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK3787 is a specific and irreversible PPAR(delta) inhibitor(pIC50= 6.6). purity: 98%