Learn More
Aviva Systems Biology Corp GSK3B Antibody - C-terminal region (ARP54644_P050) | 100uL
SDP
Catalog No. 5001461273
Encompass_Preferred
This is a rabbit polyclonal antibody against GSK3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GSK3B. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61273 Supplier Aviva Systems Biology Corp Supplier No. ARP54644_P050
This is a rabbit polyclonal antibody against GSK3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GSK3B. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish