Learn More
Medchemexpress LLC HY-101027A 100mg Medchemexpress, GSK4028 CAS:2079886-19-2 Purity:>98%
SDP
Catalog No. 501932014
Encompass_Preferred
Medchemexpress, HY-101027A 100mg GSK4028 CAS:2079886-19-2 GSK4028 is the enantiomeric negative control of GSK4027, which is a PCAF/GCN5 bromodomain chemical probe, the pIC 50 of GSK4028 is 4.9 in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2014 Supplier Medchemexpress LLC Supplier No. HY101027A100MG
Medchemexpress, HY-101027A 100mg GSK4028 CAS:2079886-19-2 GSK4028 is the enantiomeric negative control of GSK4027, which is a PCAF/GCN5 bromodomain chemical probe, the pIC 50 of GSK4028 is 4.9 in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.