Learn More
Medchemexpress LLC GSK4112 (SR6452) | 1216744-19-2 | 99.9% | C18H21ClN2O4S | 100 MG
SDP
Catalog No. 5000354069
Encompass_Preferred
GSK4112, also known as SR6452, is identified as a Rev-erbα agonist with an EC50 value of 0.4 μM. It serves as a valuable chemical tool for investigating the role of Rev-erbα in various biological processes, including transcriptional repression, the regulation of circadian rhythms, and metabolic pathways.
  • Interacts with Rev-erbα with an EC50 of 0.4 μM.
  • Represses the expression of bmal1 and target genes associated with gluconeogenesis, recruits HDAC3, and modulates Rev-erbα's effect on hepatic gene expression oscillation.
  • Reduces glucose output in murine hepatocytes.
  • Attenuates Fas-induced hepatic damage in male C57BL/6 mice.
Catalog No. 50-003-54069 Supplier Medchemexpress LLC Supplier No. HY14414100MG
GSK4112, also known as SR6452, is identified as a Rev-erbα agonist with an EC50 value of 0.4 μM. It serves as a valuable chemical tool for investigating the role of Rev-erbα in various biological processes, including transcriptional repression, the regulation of circadian rhythms, and metabolic pathways.
  • Interacts with Rev-erbα with an EC50 of 0.4 μM.
  • Represses the expression of bmal1 and target genes associated with gluconeogenesis, recruits HDAC3, and modulates Rev-erbα's effect on hepatic gene expression oscillation.
  • Reduces glucose output in murine hepatocytes.
  • Attenuates Fas-induced hepatic damage in male C57BL/6 mice.