Learn More
Apexbio Technology LLC GSK461364, 50mg. Cas: 929095-18-1 MFCD: MFCD12755419. Plk1 inhibitor
SDP
Supplier:  Apexbio Technology LLC A844150
Encompass_Preferred
GSK461364 is a potent and reversible ATP competitive Plk1 inhibitor. Polo-like kinases (Plk) are a family of serine threonine kinases that are critical regulators of DNA damage response and cell cycle progression. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5143