Learn More
Selleck Chemical LLC GSK461364 10mg 929095-18-1
SDP
Supplier:  Selleck Chemical LLC S219310MG
Encompass_Preferred
GSK461364 inhibits purified Plk1 with Ki of 2.2 nM in a cell-free assay. It is more than 1000-fold selective against Plk2/3. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-859-9