Learn More
Medchemexpress LLC HY-116761 10mg Medchemexpress, GSK467 CAS:1628332-52-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11676110MG
Encompass_Preferred
Medchemexpress, HY-116761 10mg GSK467 CAS:1628332-52-4 GSK467 is a cell penetrant and selective KDM5B (JARID1B or PLU1) inhibitor with a Ki of 10 nM, shows 180-fold selectivity for KDM4C and no measurable inhibitory effects toward KDM6 or other Jumonji family members. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9438