Learn More
Medchemexpress LLC HY-100514 25mg , GSK484 (hydrochloride) CAS:1652591-81-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10051425MG
Encompass_Preferred
HY-100514 25mg GSK484 (hydrochloride) CAS: 1652591-81-5 GSK484 hydrochloride is a selective and reversible peptidylarginine deiminase 4 (PAD4) inhibitor. GSK484 hydrochloride demonstrates high affinity binding to PAD4 with IC50s of 50 nM in the absence of Calcium. In the presence of 2 mM Calcium, notably lower potency (250 nM) is observed. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9862