Learn More
Selleck Chemical LLC GSK484 HCl 5mg
SDP
Supplier:  Selleck Chemical LLC S78035MG
Encompass_Preferred
GSK484 HCl, a benzoimidazole derivative, is a selective and reversible inhibitor of peptidylarginine deiminase 4 (PAD4) with IC50 of 50 nM in the absence of Calcium. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3309