Learn More
Cayman Chemical GSK484hydrochlorIde 10mg
SDP
Catalog No. 502968321
Encompass_Preferred
A reversible inhibitor of PAD4 (IC50 = 50 nM) that binds to the low-calcium form of the enzyme; blocks the citrullination of PAD4 target proteins in human neutrophils and inhibits the formation of NETs in both mouse and human neutrophils
Catalog No. 50-296-8321 Supplier Cayman Chemical Supplier No. 1748810 MG
A reversible inhibitor of PAD4 (IC50 = 50 nM) that binds to the low-calcium form of the enzyme; blocks the citrullination of PAD4 target proteins in human neutrophils and inhibits the formation of NETs in both mouse and human neutrophils