Learn More
Medchemexpress LLC HY-12856 100mg , GSK503 CAS:1346572-63-1 Purity:98%
SDP
Catalog No. 501493430
Encompass_Preferred
Medchemexpress, HY-12856 100mg GSK503 CAS:1346572-63-1 Formula:C31H38N6O2 Ki: 3 to 27 nM (EZH2) Purity:98% GSK503 is a potent and specific inhibitor of EZH2 methyltransferase with K i app values of 3 to 27 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3430 Supplier Medchemexpress LLC Supplier No. HY12856100MG
Medchemexpress, HY-12856 100mg GSK503 CAS:1346572-63-1 Formula:C31H38N6O2 Ki: 3 to 27 nM (EZH2) Purity:98% GSK503 is a potent and specific inhibitor of EZH2 methyltransferase with K i app values of 3 to 27 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.