Learn More
Abcam GSK503, EZH2 methyltransferase inhibitor, 50MG
Catalog No. 502683345
Encompass_Preferred
MW 526.7 Da. Potent (IC₅₀ = nM; Ki = 3 nM) and specific EZH2 methyltransferase inhibitor. Selective against EZH1 (IC₅₀ = 633 nM). At least 4000-fold selective over a panel of 20 other human methyltransferases. Highly selective against a range of human histone deacetylases, GPCRs, kinases and ion channels. Reduces cellular H3K27me3 levels. Inhibits growth of diffuse large B-cell lymphoma cells. Inhinits melanoma growth and metastasis in mouse models.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-3345 Supplier Abcam Supplier No. AB26981950MG
MW 526.7 Da. Potent (IC₅₀ = nM; Ki = 3 nM) and specific EZH2 methyltransferase inhibitor. Selective against EZH1 (IC₅₀ = 633 nM). At least 4000-fold selective over a panel of 20 other human methyltransferases. Highly selective against a range of human histone deacetylases, GPCRs, kinases and ion channels. Reduces cellular H3K27me3 levels. Inhibits growth of diffuse large B-cell lymphoma cells. Inhinits melanoma growth and metastasis in mouse models.
The product is subject to the following: Abcam Restricted Use Statement