Learn More
Medchemexpress LLC HY-114492 10mg Medchemexpress, GSK547 CAS:2226735-55-1 Purity:>98%
SDP
Catalog No. 501969364
Encompass_Preferred
Medchemexpress, HY-114492 10mg GSK547 CAS:2226735-55-1 GSK547 (GSK'547) is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1), inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9364 Supplier Medchemexpress LLC Supplier No. HY11449210MG
Medchemexpress, HY-114492 10mg GSK547 CAS:2226735-55-1 GSK547 (GSK'547) is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1), inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.