Learn More
Medchemexpress LLC HY-114492 5mg Medchemexpress, GSK547 CAS:2226735-55-1 Purity:>98%
SDP
Catalog No. 501932494
Encompass_Preferred
Medchemexpress, HY-114492 5mg GSK547 CAS:2226735-55-1 GSK547 (GSK'547) is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1) , inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2494 Supplier Medchemexpress LLC Supplier No. HY1144925MG
Medchemexpress, HY-114492 5mg GSK547 CAS:2226735-55-1 GSK547 (GSK'547) is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1) , inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.